accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNFFF

CAS RN 66548-69-4

IUPAC Name: 3-methyl-6-[3-(trifluoromethyl)phenyl]-[1,2,4]triazolo[4,3-b]pyridazine
Synonyms: 3-methyl-6-[3-(trifluoromethyl)phenyl]-[1,2,4]triazolo[4,3-b]pyridazine
Molecular Weight (g/mol): 278.24
Molecular Formula: C13H9F3N4
InChi Key: GUOQUXNJZHGPQF-UHFFFAOYSA-N
SMILES: CC1=NN=C2C=CC(=NN12)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results