Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CL 218872, Tocris Bioscience™

Catalog No. 170910
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-091-0 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-091-0 Supplier R&D Systems Supplier No. 1709/10
This item is not returnable. View return policy

Benzodiazepine agonist

Benzodiazepine agonist displaying selectivity for α1 subunit-containing GABAA receptors (Ki values are 130, 1820, 1530, > 10000, 490 and > 10000 nM for α1, α2, α3, α4, α5 and α6-subunit containing receptors respectively). Orally active anxiolytic and anticonvulsant in vivo.

Specifications

CAS 66548-69-4
Molecular Formula C13H9F3N4
Quantity 10 mg
Synonym 3-methyl-6-3-trifluoromethyl phenyl-1,2,4 triazolo 4,3-b pyridazine, chembl13662, 3-methyl-6-3-trifluoromethyl phenyl-1,2,4-triazolo 4,2-b pyridazine, 1,2,4-triazolo 4,3-b pyridazine, 3-methyl-6-3-trifluoromethyl phenyl, 3-methyl-6-3-trifluoromethyl phenyl-1,2,4-triazolo-4,3-b pyridazine, 3-methyl-6--3-trifluoromethyl phenyl-1,2,4-triazolo 4,3-b pyridazine, tocris-1709
InChI Key GUOQUXNJZHGPQF-UHFFFAOYSA-N
SMILES CC1=NN=C2N1N=C(C=C2)C3=CC(=CC=C3)C(F)(F)F
IUPAC Name 3-methyl-6-[3-(trifluoromethyl)phenyl]-[1,2,4]triazolo[4,3-b]pyridazine
Molecular Weight (g/mol) 278.238
PubChem CID 107950
Formula Weight 278.24
Percent Purity 98%
Chemical Name or Material CL 218872
Show More Show Less
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.