accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHSOOClClCl

CAS RN 6655-72-7

IUPAC Name: 2,4,5-trichlorobenzene-1-sulfonohydrazide
Synonyms: 2,4,5-trichlorobenzenesulfonohydrazide
Molecular Weight (g/mol): 275.53
Molecular Formula: C6H5Cl3N2O2S
InChi Key: PDXFRYXZDLBGGJ-UHFFFAOYSA-N
SMILES: NNS(=O)(=O)C1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results