Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichlorobenzenesulfonyl Hydrazide 98%, Thermo Scientific™

Catalog No. AC151140050
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC151140050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC151140050 Supplier Thermo Scientific Chemicals Supplier No. 151140050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6655-72-7
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C6H5Cl3N2O2S
Linear Formula Cl3C6H2SO2NHNH2
Quantity 5g
Synonym 2,4,5-trichlorobenzenesulfonyl hydrazide, benzenesulfonic acid, 2,4,5-trichloro-, hydrazide, 2,4,5-trichlorobenzenesulphonohydrazide, benzenesulfonic acid,2,4,5-trichloro-, hydrazide, 2,5-trichlorobenzenesulfonyl hydrazide, hydrazino 2,4,5-trichlorophenyl sulfone, benzenesulfonic acid,4,5-trichloro-, hydrazide
InChI Key PDXFRYXZDLBGGJ-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)NN
IUPAC Name 2,4,5-trichlorobenzenesulfonohydrazide
Molecular Weight (g/mol) 275.528
PubChem CID 81167
Formula Weight 275.53
Percent Purity ≥97.5%
Chemical Name or Material 2, 4, 5-Trichlorobenzenesulfonyl hydrazide, 98%
Show More Show Less
EINECSNumber 229-680-9
RTECSNumber DB8200000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.