accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66556-73-8

IUPAC Name: (2S)-2-[(2R)-2-[(2S)-2-[(2R)-2-[(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanamido]-4-methylpentanamido]-3-phenylpropanamido]-4-methylpentanamido]-3-phenylpropanoic acid
Synonyms: (2S)-2-[(2R)-2-[(2S)-2-[(2R)-2-[(2S)-2-[(tert-butoxycarbonyl)amino]-3-phenylpropanamido]-4-methylpentanamido]-3-phenylpropanamido]-4-methylpentanamido]-3-phenylpropanoic acid
Molecular Weight (g/mol): 785.98
Molecular Formula: C44H59N5O8
InChi Key: NGNZQSPFQJCBJQ-DWCHZDDLSA-N
SMILES: CC(C)C[C@@H](NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)OC(C)(C)C)C(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results