Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-t-BOC-Phe-Leu-Phe-Leu-Phe, 97.5%, MP Biomedicals™

Catalog No. ICN15276025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15276025 25 mg
ICN15276005 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15276025 Supplier MP Biomedicals, Inc Supplier No. 0215276025
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66556-73-8
Molecular Formula C44H59N5O8
Molecular Weight (g/mol) 785.98
InChI Key NGNZQSPFQJCBJQ-DWCHZDDLSA-N
Synonym boc-phe-d-leu-phe-d-leu-phe-oh, bplplp, butyloxycarbonyl-phenylalanyl-leucyl-phenylalanyl-leucyl-phenylalanine, n-t-boc-phe-leu-phe-leu-phe, n-t-boc-phe-dleu-phe-dleu-phe, butyloxycarbonyl-phe-leu-phe-leu-phe, n-tertiary-butyloxycarbonyl-phenylalanyl-leucyl-phenylalanyl-leucyl-phenylalanine, 2s-2-2r-4-methyl-2-2s-2-2r-4-methyl-2-2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenyl-propanoyl amino pentanoyl amino-3-phenyl-propanoyl amino pentanoyl amino-3-phenyl-propanoic acid, 2s-2-2r-4-methyl-2-2s-2-2r-4-methyl-2-2s-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoyl amino pentanoyl amino-3-phenylpropanoyl amino pentanoyl amino-3-phenylpropanoic acid, 2s-2-2r-4-methyl-2-2s-2-2r-4-methyl-2-2s-2-2-methylpropan-2-yl oxy-oxomethyl amino-1-oxo-3-phenylpropyl amino-1-oxopentyl amino-1-oxo-3-phenylpropyl amino-1-oxopentyl amino-3-phenylpropanoic acid
PubChem CID 125628

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.