accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FFFNOO

CAS RN 6656-49-1

IUPAC Name: 2-methyl-1-nitro-3-(trifluoromethyl)benzene
Synonyms: 2-methyl-1-nitro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: KQUQBPVYIURTNZ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1C(F)(F)F)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)