Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitrobenzotrifluoride 97.0+%, TCI America™
SDP

Catalog No. M12755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12755G 5 g
M127525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12755G Supplier TCI America Supplier No. M12755G

Chemical Identifiers

CAS 6656-49-1
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD00042322
InChI Key KQUQBPVYIURTNZ-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzotrifluoride, 2-nitro-6-trifluoromethyltoluene, 2-methyl-1-nitro-3-trifluoromethyl benzene, 2-nitro-6-trifluoromethyl toluene, 2-methyl-3-trifluoromethyl nitrobenzene, benzene, 2-methyl-1-nitro-3-trifluoromethyl, alpha,alpha,alpha-trifluoro-3-nitro-o-xylene, 3-trifluoromethyl-2-methyl-1-nitrobenzene, pubchem4816, 2-methyl-3-nitro-benzotrifluoride
PubChem CID 81171
IUPAC Name 2-methyl-1-nitro-3-(trifluoromethyl)benzene
SMILES CC1=C(C=CC=C1C(F)(F)F)[N+]([O-])=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 205.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-nitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.