CAS RN 666837-93-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ SU9516
A cell-permeable 3-substituted indolinone compound that displays anti-proliferative and proapoptotic properties in tumor cells.