accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHNBrHNO(E/Z)

CAS RN 667463-85-6

IUPAC Name: {6'-bromo-2'-oxo-1',2'-dihydro-[2,3'-biindolyliden]-3-yl}amino acetate
Synonyms: {6'-bromo-2'-oxo-1'H-[2,3'-biindolyliden]-3-yl}amino acetate
Molecular Weight (g/mol): 398.22
Molecular Formula: C18H12BrN3O3
InChi Key: ORWSYHWTBSHFIV-UHFFFAOYSA-N
SMILES: CC(=O)ONC1=C2C=CC=CC2=NC1=C1C(=O)NC2=C1C=CC(Br)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results