accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 668-45-1

IUPAC Name: 2-chloro-6-fluorobenzonitrile
Synonyms: 2-chloro-6-fluorobenzonitrile
Molecular Weight (g/mol): 155.56
Molecular Formula: C7H3ClFN
InChi Key: XPTAYRHLHAFUOS-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Cl)=C1C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results