accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 66990-30-5

IUPAC Name: tricosa-10,12-diynoic acid
Synonyms: tricosa-10,12-diynoic acid
Molecular Weight (g/mol): 346.56
Molecular Formula: C23H38O2
InChi Key: DIEDVCMBPCRJFQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC#CC#CCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)