Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,12-Tricosadiynoic Acid 98.0+%, TCI America™
SDP

Catalog No. T12001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12001G Supplier TCI America Supplier No. T12001G

missing translation for 'validMainframeMsg'

Specifications

CAS 66990-30-5
Melting Point 59°C
Color White-Yellow
Molecular Formula C23H38O2
MDL Number MFCD00041683
Quantity 1 g
Synonym 10,12-tricosadiynoic acid, 10,12-tricosadiynoicacid, acmc-209nx8, 10,12-tricosadiynoic acid gc
InChI Key DIEDVCMBPCRJFQ-UHFFFAOYSA-N
SMILES CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
IUPAC Name tricosa-10,12-diynoic acid
Molecular Weight (g/mol) 346.555
PubChem CID 538457
Formula Weight 346.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 10,12-Tricosadiynoic Acid
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.