accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66990-34-9

IUPAC Name: nonacosa-10,12-diynoic acid
Synonyms: nonacosa-10,12-diynoic acid
Molecular Weight (g/mol): 430.72
Molecular Formula: C29H50O2
InChi Key: LQESNAREESPTNY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results