Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,12-Nonacosadiynoic Acid 97.0+%, TCI America™
SDP

Catalog No. N0514100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0514100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0514100MG Supplier TCI America Supplier No. N0514100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66990-34-9
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00060119
InChI Key LQESNAREESPTNY-UHFFFAOYSA-N
PubChem CID 4097613
IUPAC Name nonacosa-10,12-diynoic acid
SMILES CCCCCCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O

Specifications

Melting Point 72°C
Quantity 100 mg
Formula Weight 430.72
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 10,12-Nonacosadiynoic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.