accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SCH3OO

CAS RN 67-71-0

IUPAC Name: methanesulfonylmethane
Synonyms: methylsulfonylmethane lignisul msm msm - methylsulfonylmethane 1,1'-sulfonylbis-methane MSM methane, 1,1'-sulfonylbis- dimethyl sulfone (substance) dimethyl sulfone, 13C-labeled 13C-labeled dimethyl sulfone opti msm
Molecular Weight (g/mol): 94.13
Molecular Formula: C2H6O2S
InChi Key: HHVIBTZHLRERCL-UHFFFAOYSA-N
SMILES: CS(C)(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (5)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
  • (2)
Physical Form 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (5)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)