Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylsulfonylmethane, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111015141 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5141 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5141 Supplier MilliporeSigma Supelco Supplier No. PHR13461G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylsulfonylmethane is the oxidized form of dimethyl sulfoxide, generally occurring in green plants, fruits and vegetables. Methylsulfonylmethane can find applications as a solvent to dissolve a wide range of solutes and is also generally used for high temperature reactions. It exhibits pharmacological properties and is reported to reduce peripheral pain, inflammation and arthritis condition.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-71-0
Physical Form Neat
Linear Formula (CH3)2 SO2
MDL Number MFCD00007566
Melting Point 107°C to 109°C
Boiling Point 238°C
Synonym Dimethyl sulfone; Methyl sulfone
Quantity 1 g
Formula Weight 94.13
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.