CAS RN 67-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydrotachysterol, ≥98%, MilliporeSigma™ Supelco™
Dihydrotachysterol is a vitamin D analog in the reduced form, which has implications in the studies of calcemic and cell differentiating action of the hormone, 1α,25-dihydroxyvitamin D3. It has been indicated as a medication for the treatment of hypoparathyroidism and renal osteodystrophy.