Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dihydrotachysterol, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111018598 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8598 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8598 Supplier MilliporeSigma Supelco Supplier No. D925750MG
Add to Cart
Add to Cart

Dihydrotachysterol is a vitamin D analog in the reduced form, which has implications in the studies of calcemic and cell differentiating action of the hormone, 1α,25-dihydroxyvitamin D3. It has been indicated as a medication for the treatment of hypoparathyroidism and renal osteodystrophy.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care, Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-96-9
Physical Form Neat
Linear Formula C28H46 O
Synonym 9,10-Secoergosta-5,7,22-trien-3 beta-ol; DHT2
Type Analytical Standard
Quantity 50 mg
Formula Weight 398.66
Percent Purity ≥98%
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.