accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3OH

CAS RN 6702-50-7

IUPAC Name: methyl 3-hydroxy-4-methoxybenzoate
Synonyms: methyl 3-hydroxy-4-methoxybenzoate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: QXOXUEFXRSIYSW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=C(OC)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)