CAS RN 6719-02-4
IUPAC Name:
2-(1H-pyrrol-1-yl)ethan-1-ol
Synonyms:
2-(pyrrol-1-yl)ethanol
Molecular Weight (g/mol):
111.14
Molecular Formula:
C6H9NO
InChi Key:
ZIOLCZCJJJNOEJ-UHFFFAOYSA-N
SMILES:
OCCN1C=CC=C1
1
–
1
of
1
results
1
1-(2-Hydroxyethyl)pyrrole 99.0+%, TCI America™
CAS: 6719-02-4 Molecular Formula: C6H9NO Molecular Weight (g/mol): 111.14 MDL Number: MFCD00191451 InChI Key: ZIOLCZCJJJNOEJ-UHFFFAOYSA-N PubChem CID: 10975438 IUPAC Name: 2-pyrrol-1-ylethanol SMILES: C1=CN(C=C1)CCO