accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNH2O

CAS RN 6719-21-7

IUPAC Name: 2-amino-2-cyanoacetamide
Synonyms: 2-amino-2-cyanoacetamide
Molecular Weight (g/mol): 99.09
Molecular Formula: C3H5N3O
InChi Key: JRWAUKYINYWSTA-UHFFFAOYNA-N
SMILES: NC(C#N)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)