accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67192-42-1

CH3NOOFFF

CAS RN 67192-42-1

IUPAC Name: 2-methyl-1-nitro-4-(trifluoromethyl)benzene
Synonyms: 2-methyl-1-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: PCCXJSOGGOWRJC-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1[N+]([O-])=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results