Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-4-nitrobenzotrifluoride, 97%, Thermo Scientific™

Catalog No. AC446460010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446460010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC446460010 Supplier Thermo Scientific Chemicals Supplier No. 446460010

Chemical Identifiers

CAS 67192-42-1
InChI Key PCCXJSOGGOWRJC-UHFFFAOYSA-N
Synonym 3-methyl-4-nitrobenzotrifluoride, 2-methyl-1-nitro-4-trifluoromethyl benzene, 4-trifluoromethyl-2-methyl-1-nitrobenzene, 2-methyl-1-nitro-4-trifluoromethyl-benzene, 3-methyl-4-nitrobenzotrifluoroide, 1-nitro-2-methyl-4-trifluoromethyl benzene, benzene, 2-methyl-1-nitro-4-trifluoromethyl
PubChem CID 144223
IUPAC Name 2-methyl-1-nitro-4-(trifluoromethyl)benzene
SMILES CC1=C(C=CC(=C1)C(F)(F)F)[N+](=O)[O-]

Specifications

Boiling Point 95.0°C (13.0 Torr)
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H6F3NO2
Quantity 1 g
Molecular Weight (g/mol) 205.13
Formula Weight 205.13
Percent Purity 97%
Chemical Name or Material 3-Methyl-4-nitrobenzotrifluoride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.