accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 672-45-7

IUPAC Name: 6-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-(trifluoromethyl)-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 180.09
Molecular Formula: C5H3F3N2O2
InChi Key: IROWWTVZNHKLLE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)NC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results