accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClI

CAS RN 672-57-1

IUPAC Name: 1-chloro-2-iodo-4-(trifluoromethyl)benzene
Synonyms: 1-chloro-2-iodo-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 306.45
Molecular Formula: C7H3ClF3I
InChi Key: SSLWFPKNYZEOTH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results