Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-iodobenzotrifluoride 99%, Thermo Scientific™

Catalog No. AC249471000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC249471000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC249471000 Supplier Thermo Scientific Chemicals Supplier No. 249471000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 672-57-1
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C7H3ClF3I
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Linear Formula CF3C6H3ClI
Quantity 100g
MDL Number MFCD00001037
Synonym 1-chloro-2-iodo-4-trifluoromethyl benzene, 4-chloro-3-iodobenzotrifluoride, 3-iodo-4-chlorobenzotrifluoride, benzene, 1-chloro-2-iodo-4-trifluoromethyl, 4-chloro-3-iodobenzo trifluoride, pubchem2714, attercop-chm at113459, 1-chloro-2-iodo-4-trifluoromethyl-benzene, 1-chloro-2-iodo-4-trifluoromethyl benzene #
InChI Key SSLWFPKNYZEOTH-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(F)(F)F)I)Cl
IUPAC Name 1-chloro-2-iodo-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 306.451
PubChem CID 69596
Formula Weight 306.44
Percent Purity ≥98.5%
Chemical Name or Material 4-Chloro-3-iodobenzotrifluoride, 99%
Show More Show Less
EINECSNumber 211-593-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.