accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNHOClOH

CAS RN 67206-28-4

IUPAC Name: 2-(2-chloroacetamido)-3-hydroxypropanoic acid
Synonyms: 2-(2-chloroacetamido)-3-hydroxypropanoic acid
Molecular Weight (g/mol): 181.57
Molecular Formula: C5H8ClNO4
InChi Key: SGCJRDDJGFXEAB-UHFFFAOYNA-N
SMILES: OCC(NC(=O)CCl)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results