Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chloroacetyl-DL-serine 95.0+%, TCI America™
SDP

Catalog No. C01041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C01041G Supplier TCI America Supplier No. C01041G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 67206-28-4
Molecular Formula C5H8ClNO4
Molecular Weight (g/mol) 181.57
MDL Number MFCD00020416
InChI Key SGCJRDDJGFXEAB-UHFFFAOYNA-N
PubChem CID 294515
IUPAC Name 2-(2-chloroacetamido)-3-hydroxypropanoic acid
SMILES OCC(NC(=O)CCl)C(O)=O

Specifications

Melting Point 122°C
Quantity 1 g
Formula Weight 181.57
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-DL-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.