CAS RN 67245-85-6
IUPAC Name:
1,3-bis[(4-nitrophenyl)methyl] propanedioate
Synonyms:
1,3-bis[(4-nitrophenyl)methyl] propanedioate
Molecular Weight (g/mol):
374.31
Molecular Formula:
C17H14N2O8
InChi Key:
RBVOYZBIDZCDBO-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(COC(=O)CC(=O)OCC2=CC=C(C=C2)[N+]([O-])=O)C=C1
1
–
1
of
1
results
1
Bis(4-nitrobenzyl) Malonate 98.0+%, TCI America™
CAS: 67245-85-6 Molecular Formula: C17H14N2O8 Molecular Weight (g/mol): 374.305 MDL Number: MFCD00191671 InChI Key: RBVOYZBIDZCDBO-UHFFFAOYSA-N Synonym: Malonic Acid Bis(4-nitrobenzyl) Ester PubChem CID: 313754 IUPAC Name: bis[(4-nitrophenyl)methyl] propanedioate SMILES: C1=CC(=CC=C1COC(=O)CC(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]