Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-nitrobenzyl) Malonate 98.0+%, TCI America™
SDP

Catalog No. M11815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11815G Supplier TCI America Supplier No. M11815G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67245-85-6
Molecular Formula C17H14N2O8
Molecular Weight (g/mol) 374.305
MDL Number MFCD00191671
InChI Key RBVOYZBIDZCDBO-UHFFFAOYSA-N
Synonym Malonic Acid Bis(4-nitrobenzyl) Ester
PubChem CID 313754
IUPAC Name bis[(4-nitrophenyl)methyl] propanedioate
SMILES C1=CC(=CC=C1COC(=O)CC(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Formula Weight 374.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrobenzyl) Malonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.