accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67254-28-8

IUPAC Name: [2,3-bis(octadec-9-enoyloxy)propoxy](2,3-dihydroxypropoxy)phosphinic acid sodium
Synonyms: 2,3-bis(octadec-9-enoyloxy)propoxy(2,3-dihydroxypropoxy)phosphinic acid sodium
Molecular Weight (g/mol): 798.05
Molecular Formula: C42H79NaO10P
InChi Key: YBQBWNNQFRRICZ-UHFFFAOYNA-N
SMILES: [Na].CCCCCCCCC=CCCCCCCCC(=O)OCC(COP(O)(=O)OCC(O)CO)OC(=O)CCCCCCCC=CCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results