Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. D5318250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5318250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D5318250MG Supplier TCI America Supplier No. D5318250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67254-28-8
Molecular Formula C42H79NaO10P
Molecular Weight (g/mol) 798.05
MDL Number MFCD00070069
InChI Key YBQBWNNQFRRICZ-UHFFFAOYNA-N
Synonym Sodium (R)-2,3-Bis(oleoyloxy)propyl (2,3-Dihydroxypropyl) Phosphate, Dioleoylphosphatidylglycerol Sodium Salt, DOPG Sodium Salt
PubChem CID 131857300
IUPAC Name [2,3-bis(octadec-9-enoyloxy)propoxy](2,3-dihydroxypropoxy)phosphinic acid sodium
SMILES [Na].CCCCCCCCC=CCCCCCCCC(=O)OCC(COP(O)(=O)OCC(O)CO)OC(=O)CCCCCCCC=CCCCCCCCC

Specifications

Color White
Quantity 250 mg
Formula Weight 797.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.