accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPFeCoClCl

CAS RN 67292-36-8

IUPAC Name: [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride
Synonyms: —
Molecular Weight (g/mol): 684.22
Molecular Formula: C34H28Cl2CoFeP2
InChi Key: —
SMILES: [Fe].Cl[Co]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)