Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) Dichloride 98.0+%, TCI America™
SDP

Catalog No. B33745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33745G 5 g
B33741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33745G Supplier TCI America Supplier No. B33745G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67292-36-8
Molecular Formula C34H28Cl2CoFeP2
Molecular Weight (g/mol) 684.22
MDL Number MFCD03427161
IUPAC Name [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride
SMILES [Fe].Cl[Co]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C
Color Green-Yellow
Quantity 5 g
Synonym [1,1′-Bis(diphenylphosphino)ferrocene]dichlorocobalt(II), Dichloro[1,1′-bis(diphenylphosphino)ferrocene]cobalt(II)
Formula Weight 684.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,1′-Bis(diphenylphosphino)ferrocene]cobalt(II) Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.