accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 673-66-5

IUPAC Name: azocan-2-one
Synonyms: 2-azacyclooctanone
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: CJYXCQLOZNIMFP-UHFFFAOYSA-N
SMILES: O=C1CCCCCCN1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)