Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Azacyclooctanone 97%, Thermo Scientific™

Catalog No. AC105080250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105080250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105080250 Supplier Thermo Scientific Chemicals Supplier No. 105080250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-66-5
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00003271
InChI Key CJYXCQLOZNIMFP-UHFFFAOYSA-N
Synonym 2-azacyclooctanone, enantholactam, omega-heptalactam, 2-perhydroazocinone, oenantholactam, 7-heptanelactam, octahydroazocin-2-one, 2 1h-azocinone, hexahydro, azacyclooctan-2-one, suberonisoxim
PubChem CID 12657
IUPAC Name azocan-2-one
SMILES O=C1CCCCCCN1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 38°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 25 g
Beilstein 21, 242
Formula Weight 127.19
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2-Azacyclooctanone, 97%
Show More Show Less
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.