accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67362-76-9

IUPAC Name: 2-butoxyethyl 4-(dimethylamino)benzoate
Synonyms: 2-butoxyethyl 4-(dimethylamino)benzoate
Molecular Weight (g/mol): 265.35
Molecular Formula: C15H23NO3
InChi Key: PAAVDLDRAZEFGW-UHFFFAOYSA-N
SMILES: CCCCOCCOC(=O)C1=CC=C(C=C1)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results