Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butoxyethyl 4-(Dimethylamino)benzoate 95.0+%, TCI America™
SDP

Catalog No. D184825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D184825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D184825G Supplier TCI America Supplier No. D184825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67362-76-9
Molecular Formula C15H23NO3
Molecular Weight (g/mol) 265.353
MDL Number MFCD00055652
InChI Key PAAVDLDRAZEFGW-UHFFFAOYSA-N
Synonym 4-(Dimethylamino)benzoic Acid 2-Butoxyethyl Ester, Ethylene Glycol Monobutyl Ether 4-(Dimethylamino)benzoate
PubChem CID 105420
IUPAC Name 2-butoxyethyl 4-(dimethylamino)benzoate
SMILES CCCCOCCOC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 265.35
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butoxyethyl 4-(Dimethylamino)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.