accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6737-24-2

IUPAC Name: (2R)-2-hydroxy-2-(prop-2-enamido)acetate
Synonyms: (2R)-2-hydroxy-2-(prop-2-enamido)acetate
Molecular Weight (g/mol): 144.11
Molecular Formula: C5H6NO4
InChi Key: NEYTXADIGVEHQD-SCSAIBSYSA-M
SMILES: O[C@@H](NC(=O)C=C)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results