Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamido glycolic acid, 98%, anhydrous, Thermo Scientific™

Catalog No. AC279950500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279950500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC279950500 Supplier Thermo Scientific Chemicals Supplier No. 279950500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6737-24-2
Molecular Formula C5H7NO4
Molecular Weight (g/mol) 145.114
InChI Key NEYTXADIGVEHQD-UHFFFAOYSA-N
Synonym acrylamidoglycolic acid, 2-acrylamidoglycolic acid, acetic acid, hydroxy 1-oxo-2-propenyl amino, hydroxy 1-oxo-2-propenyl amino acetic acid, hydroxy 1-oxoallyl amino acetic acid, glycolic acid, acrylamido-7ci,8ci, 2-hydroxy-2-1-oxo-2-propenyl amino acetic acid, acetic acid, 2-hydroxy-2-1-oxo-2-propen-1-yl amino
PubChem CID 92230
IUPAC Name 2-hydroxy-2-(prop-2-enoylamino)acetic acid
SMILES C=CC(=O)NC(C(=O)O)O

Specifications

Melting Point 123°C to 127°C
CAS Min % 0.975
Color White
CAS Max % 1
Assay Percent Range 0.98
Linear Formula CH2=CHC(O)nHCH(OH)COOH
Formula Weight 145.11
Percent Purity ≥97.5%
Quantity 50 g
Infrared Spectrum Authentic
Physical Form Crystalline Powder
Chemical Name or Material Acrylamido glycolic acid, 0.98
Show More Show Less
Hazard Category LIGHT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.