accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3H3CSiCH3CH3Cl

CAS RN 67373-56-2

IUPAC Name: chloro(2,3-dimethylbutan-2-yl)dimethylsilane
Synonyms: chloro(2,3-dimethylbutan-2-yl)dimethylsilane
Molecular Weight (g/mol): 178.78
Molecular Formula: C8H19ClSi
InChi Key: KIGALSBMRYYLFJ-UHFFFAOYSA-N
SMILES: CC(C)C(C)(C)[Si](C)(C)Cl
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)