Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(Dimethyl)Thexylsilane

Catalog No. 30244925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-244-925G 25 g
30-244-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 30-244-925G Supplier Sigma Aldrich Supplier No. 30244925G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protecting reagent for alcohols and other functional groups. Employed in the synthesis of silicon naphthocyanine and cyclodextrin derivatives, which are useful as antitumor dyes and chiral stationary phases, respectively.

Specifications

CAS 67373-56-2
Density 0.909 g/mL at 25°C
Boiling Point 55°C to 56°C (10 mmHg, lit.)
Molecular Formula C8H19ClSi
Refractive Index n20/D 1.449
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
MDL Number MFCD00012175
Quantity 25 g
Synonym Chlorodimethyl(1,1,2-trimethylpropyl)silane; Dimethyl-(2,3-dimethyl-2-butyl)chlorosilane; Dimethyl-thexylchlorosilane; Dimethylthexylsilyl chloride; TDSCl; Thexyldimethylsilyl chloride
Molecular Weight (g/mol) 178.77 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.