accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67386-38-3

IUPAC Name: hydrogen 2-phenoxyethanimidamide chloride
Synonyms: hydrogen 2-phenoxyethanimidamide chloride
Molecular Weight (g/mol): 186.64
Molecular Formula: C8H11ClN2O
InChi Key: ZLZLLGGMCNVVAS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)COC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results