accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67421-02-7

IUPAC Name: bis(λ²-ruthenium(2+)) bis(hexamethylbenzene) tetrachloride
Synonyms: bis(λ²-ruthenium(2+)) bis(hexamethylbenzene) tetrachloride
Molecular Weight (g/mol): 668.49
Molecular Formula: C24H36Cl4Ru2
InChi Key: GVCIEWAYDUQTTQ-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].CC1=C(C)C(C)=C(C)C(C)=C1C.CC1=C(C)C(C)=C(C)C(C)=C1C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results