Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Hexamethylbenzene)ruthenium(II) Dichloride Dimer 95.0+%, TCI America™
SDP

Catalog No. H10101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10101G Supplier TCI America Supplier No. H10101G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67421-02-7
Molecular Formula C24H36Cl4Ru2
Molecular Weight (g/mol) 668.492
MDL Number MFCD03701102
InChI Key GVCIEWAYDUQTTQ-UHFFFAOYSA-J
Synonym Dichloro(hexamethylbenzene)ruthenium(II) Dimer
PubChem CID 53384554
IUPAC Name 1,2,3,4,5,6-hexamethylbenzene;ruthenium(2+);tetrachloride
SMILES CC1=C(C(=C(C(=C1C)C)C)C)C.CC1=C(C(=C(C(=C1C)C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2]

Specifications

Quantity 1 g
Formula Weight 668.49
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Hexamethylbenzene)ruthenium(II) Dichloride Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.