accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67488-46-4

IUPAC Name: methyl 2,5-dihydrothiophene-3-carboxylate
Synonyms: methyl 2,5-dihydrothiophene-3-carboxylate
Molecular Weight (g/mol): 144.19
Molecular Formula: C6H8O2S
InChi Key: RDHSPSSFHQGPDS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CCSC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results