accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67503-46-2

IUPAC Name: 6-{[2-(3-hydroxyphenoxy)diazen-1-yl]oxy}-[1,1'-biphenyl]-2-ol
Synonyms: 6-{[2-(3-hydroxyphenoxy)diazen-1-yl]oxy}-[1,1'-biphenyl]-2-ol
Molecular Weight (g/mol): 322.32
Molecular Formula: C18H14N2O4
InChi Key: KVQRGKQWGHNYJE-UHFFFAOYSA-N
SMILES: OC1=CC(ON=NOC2=CC=CC(O)=C2C2=CC=CC=C2)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results