CAS RN 67515-56-4
IUPAC Name:
4-fluoro-3-(trifluoromethyl)benzoyl chloride
Synonyms:
4-fluoro-3-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol):
226.55
Molecular Formula:
C8H3ClF4O
InChi Key:
BUDISZQHCHGLJW-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1C(F)(F)F)C(Cl)=O
1
–
1
of
1
results
1
4-Fluoro-3-(trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™
CAS: 67515-56-4 Molecular Formula: C8H3ClF4O Molecular Weight (g/mol): 226.55 MDL Number: MFCD00061159 InChI Key: BUDISZQHCHGLJW-UHFFFAOYSA-N PubChem CID: 144253 IUPAC Name: 4-fluoro-3-(trifluoromethyl)benzoyl chloride SMILES: FC1=CC=C(C=C1C(F)(F)F)C(Cl)=O