Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-3-(trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. F06595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06595G 5 g
F06591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06595G Supplier TCI America Supplier No. F06595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67515-56-4
Molecular Formula C8H3ClF4O
Molecular Weight (g/mol) 226.55
MDL Number MFCD00061159
InChI Key BUDISZQHCHGLJW-UHFFFAOYSA-N
PubChem CID 144253
IUPAC Name 4-fluoro-3-(trifluoromethyl)benzoyl chloride
SMILES FC1=CC=C(C=C1C(F)(F)F)C(Cl)=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 3265
Synonym 4-fluoro-3-trifluoromethyl benzoyl chloride, 4-fluoro-3-trifluoromethyl benzoylchloride, 4-fluoro-3-trifluoromethyl-benzoyl chloride, benzoyl chloride,4-fluoro-3-trifluoromethyl, benzoyl chloride, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-m-toluoyl chloride, acmc-1b7wd, 4-fluoro-5-trifluoromethylbenzoyl chloride, 5-chlorocarbonyl-2-fluorobenzotrifluoride
Formula Weight 226.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.